Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGGCGGGATCTCCCGGCAGGTA[A/G]AGGTAACCCCGTGGGGTGGGGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611226 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARMC3 - armadillo repeat containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282745.1 | Intron | NP_001269674.1 | ||||
NM_001282746.1 | Intron | NP_001269675.1 | ||||
NM_001282747.1 | Intron | NP_001269676.1 | ||||
NM_173081.4 | Intron | NP_775104.2 | ||||
XM_005252380.2 | Intron | XP_005252437.1 | ||||
XM_005252381.2 | Intron | XP_005252438.1 | ||||
XM_011519350.2 | Intron | XP_011517652.1 | ||||
XM_011519351.2 | Intron | XP_011517653.1 | ||||
XM_017015831.1 | Intron | XP_016871320.1 | ||||
XM_017015832.1 | Intron | XP_016871321.1 | ||||
XM_017015833.1 | Intron | XP_016871322.1 |