Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGGCAGAGACTTCCTCCTGGA[A/C]TGAATTCAAGGGAGGAGCACCCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 188300 | ||||||||||||||||||||
Literature Links: |
AFMID PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AFMID - arylformamidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010982.4 | Intron | NP_001010982.2 | ||||
NM_001145526.2 | Intron | NP_001138998.1 | ||||
XM_011524326.2 | Intron | XP_011522628.1 | ||||
XM_011524327.2 | Intron | XP_011522629.1 | ||||
XM_011524328.2 | Intron | XP_011522630.1 | ||||
XM_011524329.2 | Intron | XP_011522631.1 | ||||
XM_011524330.2 | Intron | XP_011522632.1 | ||||
XM_011524331.2 | Intron | XP_011522633.1 | ||||
XM_011524332.2 | Intron | XP_011522634.1 | ||||
XM_011524333.2 | Intron | XP_011522635.1 | ||||
XM_011524334.2 | Intron | XP_011522636.1 | ||||
XM_011524335.2 | Intron | XP_011522637.1 | ||||
XM_011524336.2 | Intron | XP_011522638.1 | ||||
XM_011524337.2 | Intron | XP_011522639.1 |
TK1 - thymidine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003258.4 | Intron | NP_003249.3 | ||||
XM_017024992.1 | Intron | XP_016880481.1 |