Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCTGGAGATTTTGGTGTTCAGC[A/G]TATCATATGAAGATAACACTGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603874 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANGPTL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANGPTL1 - angiopoietin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004673.3 | 2663 | UTR 3 | NP_004664.1 | |||
XM_005245577.2 | 2663 | UTR 3 | XP_005245634.1 |
RALGPS2 - Ral GEF with PH domain and SH3 binding motif 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286247.1 | 2663 | Intron | NP_001273176.1 | |||
NM_152663.4 | 2663 | Intron | NP_689876.2 | |||
XM_006711410.3 | 2663 | Intron | XP_006711473.1 | |||
XM_006711411.3 | 2663 | Intron | XP_006711474.1 | |||
XM_011509688.1 | 2663 | Intron | XP_011507990.1 | |||
XM_017001591.1 | 2663 | Intron | XP_016857080.1 |
Set Membership: |
HapMap |