Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAAGGTAGGTTTAAGATTTTTG[A/G]AGGTCTATAAACTTTGTAATAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TDRD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TDRD5 - tudor domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199085.1 | Intron | NP_001186014.1 | ||||
NM_001199089.1 | Intron | NP_001186018.1 | ||||
NM_001199091.1 | Intron | NP_001186020.1 | ||||
NM_001199092.1 | Intron | NP_001186021.1 | ||||
NM_173533.3 | Intron | NP_775804.2 | ||||
XM_005244934.1 | Intron | XP_005244991.1 | ||||
XM_005244935.4 | Intron | XP_005244992.1 | ||||
XM_017000473.1 | Intron | XP_016855962.1 |