Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGAACCACCAGATAAAAGGAA[A/C]CAATGTGGCAGATAACACCATTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300460 | ||||||||||||||||||||
Literature Links: |
PCDH19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCDH19 - protocadherin 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105243.1 | 9511 | UTR 3 | NP_001098713.1 | |||
NM_001184880.1 | 9511 | UTR 3 | NP_001171809.1 | |||
NM_020766.2 | 9511 | UTR 3 | NP_065817.2 | |||
XM_011530997.2 | 9511 | UTR 3 | XP_011529299.1 |