Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTGTGAACCTAAGAATCTCTCCT[A/C]CAAAAAAAAAAAAAAAAAAAAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613360 | ||||||||||||||||||||
Literature Links: |
DRAM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DRAM2 - DNA damage regulated autophagy modulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178454.4 | Intron | NP_848549.3 | ||||
XM_005270469.2 | Intron | XP_005270526.1 | ||||
XM_005270470.1 | Intron | XP_005270527.1 | ||||
XM_006710361.1 | Intron | XP_006710424.1 | ||||
XM_006710362.1 | Intron | XP_006710425.1 | ||||
XM_011540707.1 | Intron | XP_011539009.1 | ||||
XM_011540708.1 | Intron | XP_011539010.1 | ||||
XM_017000320.1 | Intron | XP_016855809.1 | ||||
XM_017000321.1 | Intron | XP_016855810.1 |