Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGGCTGCCGAGCCGGGACCCCC[A/G]CGCACCCCGCGCCCCGGGCGGCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610290 | ||||||||||||||||||||
Literature Links: |
GALNT12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GALNT12 - polypeptide N-acetylgalactosaminyltransferase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024642.4 | 159 | Silent Mutation | CCA,CCG | P,P 53 | NP_078918.3 | |
XM_006717287.1 | 159 | Intron | XP_006717350.1 | |||
XM_011519018.2 | 159 | Intron | XP_011517320.1 | |||
XM_011519020.1 | 159 | Intron | XP_011517322.1 | |||
XM_017015133.1 | 159 | Intron | XP_016870622.1 |