Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCTCTAAACCTTCTGAAAACTT[C/T]CCACTCAGAAAATGTGAGTTCTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 167409 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PAX2 - paired box 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000278.4 | Intron | NP_000269.3 | ||||
NM_001304569.1 | Intron | NP_001291498.1 | ||||
NM_003987.4 | Intron | NP_003978.3 | ||||
NM_003988.4 | Intron | NP_003979.2 | ||||
NM_003989.4 | Intron | NP_003980.3 | ||||
NM_003990.4 | Intron | NP_003981.3 | ||||
XM_017016281.1 | Intron | XP_016871770.1 | ||||
XM_017016282.1 | Intron | XP_016871771.1 | ||||
XM_017016283.1 | Intron | XP_016871772.1 | ||||
XM_017016284.1 | Intron | XP_016871773.1 | ||||
XM_017016285.1 | Intron | XP_016871774.1 | ||||
XM_017016286.1 | Intron | XP_016871775.1 | ||||
XM_017016287.1 | Intron | XP_016871776.1 | ||||
XM_017016288.1 | Intron | XP_016871777.1 | ||||
XM_017016289.1 | Intron | XP_016871778.1 |