Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATCGGGGGAGGACGCCTTGGGC[G/T]GGGCGTGGGTGAGAGAACCTGGGGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 139360 MIM: 604437 | |||||||||||||||||||||||
Literature Links: |
GNAI2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GNAI2 - G protein subunit alpha i2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166425.1 | Intron | NP_001159897.1 | ||||
NM_001282617.1 | Intron | NP_001269546.1 | ||||
NM_001282618.1 | Intron | NP_001269547.1 | ||||
NM_001282619.1 | Intron | NP_001269548.1 | ||||
NM_001282620.1 | Intron | NP_001269549.1 | ||||
NM_002070.3 | Intron | NP_002061.1 |
MIR5787 - microRNA 5787 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC38A3 - solute carrier family 38 member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |