Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGGATTACAGGTGAGAGCCAC[C/T]GCGTATGGCCCAAGAAAATAATTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610436 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RTTN PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RTTN - rotatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318520.1 | Intron | NP_001305449.1 | ||||
NM_173630.3 | Intron | NP_775901.3 | ||||
XM_006722434.3 | Intron | XP_006722497.1 | ||||
XM_006722435.3 | Intron | XP_006722498.1 | ||||
XM_011525902.2 | Intron | XP_011524204.1 | ||||
XM_011525903.2 | Intron | XP_011524205.1 | ||||
XM_011525904.2 | Intron | XP_011524206.1 | ||||
XM_011525905.2 | Intron | XP_011524207.1 | ||||
XM_011525906.2 | Intron | XP_011524208.1 | ||||
XM_011525907.2 | Intron | XP_011524209.1 | ||||
XM_011525908.2 | Intron | XP_011524210.1 | ||||
XM_017025693.1 | Intron | XP_016881182.1 | ||||
XM_017025694.1 | Intron | XP_016881183.1 | ||||
XM_017025695.1 | Intron | XP_016881184.1 | ||||
XM_017025696.1 | Intron | XP_016881185.1 |