Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACCTTATTTTCCTCACTGAACC[G/T]CCTTCCTTAGTTTCCTTTGCCTTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164720 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ETS1 - ETS proto-oncogene 1, transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143820.1 | 2886 | UTR 3 | NP_001137292.1 | |||
NM_001162422.1 | 2886 | UTR 3 | NP_001155894.1 | |||
NM_005238.3 | 2886 | UTR 3 | NP_005229.1 | |||
XM_011542649.2 | 2886 | UTR 3 | XP_011540951.1 | |||
XM_011542650.1 | 2886 | UTR 3 | XP_011540952.1 | |||
XM_011542651.1 | 2886 | UTR 3 | XP_011540953.1 | |||
XM_011542652.1 | 2886 | UTR 3 | XP_011540954.1 | |||
XM_017017314.1 | 2886 | UTR 3 | XP_016872803.1 | |||
XM_017017315.1 | 2886 | UTR 3 | XP_016872804.1 | |||
XM_017017316.1 | 2886 | UTR 3 | XP_016872805.1 | |||
XM_017017317.1 | 2886 | UTR 3 | XP_016872806.1 |
Set Membership: |
HapMap |