Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAGTTAGGACTATAGGCATGCA[A/C]CACCATGCCAGTCTAATTTTTAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602426 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927164 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927164 - uncharacterized LOC101927164 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NVL - nuclear VCP-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243146.1 | Intron | NP_001230075.1 | ||||
NM_001243147.1 | Intron | NP_001230076.1 | ||||
NM_002533.3 | Intron | NP_002524.2 | ||||
NM_206840.2 | Intron | NP_996671.1 | ||||
XM_011544196.1 | Intron | XP_011542498.1 | ||||
XM_011544198.1 | Intron | XP_011542500.1 | ||||
XM_011544199.1 | Intron | XP_011542501.1 | ||||
XM_011544200.1 | Intron | XP_011542502.1 | ||||
XM_017001378.1 | Intron | XP_016856867.1 | ||||
XM_017001379.1 | Intron | XP_016856868.1 | ||||
XM_017001380.1 | Intron | XP_016856869.1 | ||||
XM_017001381.1 | Intron | XP_016856870.1 | ||||
XM_017001382.1 | Intron | XP_016856871.1 | ||||
XM_017001383.1 | Intron | XP_016856872.1 | ||||
XM_017001384.1 | Intron | XP_016856873.1 | ||||
XM_017001385.1 | Intron | XP_016856874.1 | ||||
XM_017001386.1 | Intron | XP_016856875.1 |