Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCCCCGAGGCGCCCGGCTCCGA[G/T]CCCGCCTGCTGTCCGCTGGCCTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605287 | ||||||||||||||||||||
Literature Links: |
ANKMY1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKMY1 - ankyrin repeat and MYND domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DUSP28 - dual specificity phosphatase 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNPEPL1 - arginyl aminopeptidase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018226.4 | 387 | Missense Mutation | GAG,GAT | E,D 129 | NP_060696.4 | |
XM_005247036.4 | 387 | Missense Mutation | GAG,GAT | E,D 129 | XP_005247093.1 |