Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAAATAAATAAAGGCAATGTAC[A/C]CTGTTTGAGGTTTGAGCAGATGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
65 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607399 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCOLN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MCOLN2 - mucolipin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153259.3 | Intron | NP_694991.2 | ||||
XM_005270719.3 | Intron | XP_005270776.1 | ||||
XM_006710552.2 | Intron | XP_006710615.1 | ||||
XM_011541187.2 | Intron | XP_011539489.1 | ||||
XM_011541188.2 | Intron | XP_011539490.1 | ||||
XM_017000923.1 | Intron | XP_016856412.1 |
Set Membership: |
HapMap |