Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608181 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKDD1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKDD1A - ankyrin repeat and death domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPG21 - spastic paraplegia 21 (autosomal recessive, Mast syndrome) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127889.4 | Intron | NP_001121361.1 | ||||
NM_001127890.4 | Intron | NP_001121362.1 | ||||
NM_016630.6 | Intron | NP_057714.1 | ||||
XM_005254437.4 | Intron | XP_005254494.1 | ||||
XM_017022297.1 | Intron | XP_016877786.1 | ||||
XM_017022298.1 | Intron | XP_016877787.1 |