Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTCCTTTGGATGTAAAATTT[A/G]TATACAGTGAAAGGCACAAGTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603930 MIM: 608830 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPHN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPHN - gephyrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024218.1 | Intron | NP_001019389.1 | ||||
NM_020806.4 | Intron | NP_065857.1 | ||||
XM_005267254.3 | Intron | XP_005267311.1 | ||||
XM_011536340.2 | Intron | XP_011534642.1 | ||||
XM_011536342.2 | Intron | XP_011534644.1 | ||||
XM_011536343.2 | Intron | XP_011534645.1 | ||||
XM_011536344.2 | Intron | XP_011534646.1 | ||||
XM_011536345.2 | Intron | XP_011534647.1 | ||||
XM_011536346.2 | Intron | XP_011534648.1 | ||||
XM_011536347.2 | Intron | XP_011534649.1 | ||||
XM_017020913.1 | Intron | XP_016876402.1 | ||||
XM_017020914.1 | Intron | XP_016876403.1 | ||||
XM_017020915.1 | Intron | XP_016876404.1 | ||||
XM_017020916.1 | Intron | XP_016876405.1 | ||||
XM_017020917.1 | Intron | XP_016876406.1 | ||||
XM_017020918.1 | Intron | XP_016876407.1 | ||||
XM_017020919.1 | Intron | XP_016876408.1 | ||||
XM_017020920.1 | Intron | XP_016876409.1 | ||||
XM_017020921.1 | Intron | XP_016876410.1 | ||||
XM_017020922.1 | Intron | XP_016876411.1 | ||||
XM_017020923.1 | Intron | XP_016876412.1 | ||||
XM_017020924.1 | Intron | XP_016876413.1 | ||||
XM_017020925.1 | Intron | XP_016876414.1 | ||||
XM_017020926.1 | Intron | XP_016876415.1 |
RDH12 - retinol dehydrogenase 12 (all-trans/9-cis/11-cis) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152443.2 | Intron | NP_689656.2 |
Set Membership: |
HapMap |