Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAATACATATTCTTCTCGTCTGC[A/G]CACAGAACACTCAAAGACCACATTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372269 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105372269 - endogenous retrovirus group K member 8 Gag polyprotein-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011528461.2 | Intron | XP_011526763.1 |
ZNF560 - zinc finger protein 560 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152476.2 | Intron | NP_689689.2 | ||||
XM_011527696.2 | Intron | XP_011525998.1 | ||||
XM_011527697.2 | Intron | XP_011525999.1 | ||||
XM_017026327.1 | Intron | XP_016881816.1 | ||||
XM_017026328.1 | Intron | XP_016881817.1 | ||||
XM_017026329.1 | Intron | XP_016881818.1 | ||||
XM_017026330.1 | Intron | XP_016881819.1 |
Set Membership: |
HapMap |