Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTGACCTGTAGCCTTCCTGAAA[A/G]CACAAACAGTCAATTAACTCATAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610856 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD30A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD30A - ankyrin repeat domain 30A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052997.2 | Intron | NP_443723.2 | ||||
XM_011519757.2 | Intron | XP_011518059.1 | ||||
XM_011519758.2 | Intron | XP_011518060.1 | ||||
XM_017016883.1 | Intron | XP_016872372.1 |