Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCTAATCCTGAGAAATACACT[G/T]CCTCTTCCTGGCACTCAGCATTATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616065 MIM: 609951 | ||||||||||||||||||||
Literature Links: |
PIANP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIANP - PILR alpha associated neural protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244014.1 | Intron | NP_001230943.1 | ||||
NM_001244015.1 | Intron | NP_001230944.1 | ||||
NM_153685.3 | Intron | NP_710152.1 | ||||
XM_011520926.2 | Intron | XP_011519228.1 |
ZNF384 - zinc finger protein 384 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |