Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTTTGGTCATAGACATAAGAAA[C/G]CAGTCAATTACTCACAGTTTGACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616082 MIM: 603070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf4 - chromosome 12 open reading frame 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAD51AP1 - RAD51 associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130862.1 | Intron | NP_001124334.1 | ||||
NM_006479.4 | Intron | NP_006470.1 |