Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAAGTGCACCAACGTACAAAAGA[A/C]TGTAACGAGGGCTATGAGAGTTATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606055 | ||||||||||||||||||||
Literature Links: |
PCTP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCTP - phosphatidylcholine transfer protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102402.2 | Intron | NP_001095872.1 | ||||
NM_021213.3 | Intron | NP_067036.2 | ||||
XM_005257549.1 | Intron | XP_005257606.1 | ||||
XM_005257550.3 | Intron | XP_005257607.1 | ||||
XM_017024913.1 | Intron | XP_016880402.1 |