Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGTAAGTAAGGGGAAGAAGAGG[A/G]AAATGTTTGCATTTTTAAGTAAGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC59 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC59 - coiled-coil domain containing 59 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL25 - methyltransferase like 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319675.1 | Intron | NP_001306604.1 | ||||
NM_032230.2 | Intron | NP_115606.2 | ||||
XM_005269186.4 | Intron | XP_005269243.2 | ||||
XM_011538827.2 | Intron | XP_011537129.1 | ||||
XM_011538828.2 | Intron | XP_011537130.1 | ||||
XM_011538829.2 | Intron | XP_011537131.1 | ||||
XM_017020020.1 | Intron | XP_016875509.1 | ||||
XM_017020021.1 | Intron | XP_016875510.1 |
Set Membership: |
HapMap |