Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACCACTGACTTACATTCTCTTC[C/T]CCCTGGTTTCCACCTTCACCTTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612654 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCHP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCHP - trichoplein keratin filament binding | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143852.1 | Intron | NP_001137324.1 | ||||
NM_032300.4 | Intron | NP_115676.1 | ||||
XM_011538836.2 | Intron | XP_011537138.1 | ||||
XM_011538837.2 | Intron | XP_011537139.1 | ||||
XM_017020024.1 | Intron | XP_016875513.1 | ||||
XM_017020025.1 | Intron | XP_016875514.1 |