Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGAGGCAACTTCTGTGATGGTC[A/G]TAATCTGAGAAACCTCAAAAGGTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VIT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
VIT - vitrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177969.1 | Intron | NP_001171440.1 | ||||
NM_001177970.1 | Intron | NP_001171441.1 | ||||
NM_001177971.1 | Intron | NP_001171442.1 | ||||
NM_001177972.1 | Intron | NP_001171443.1 | ||||
NM_053276.3 | Intron | NP_444506.2 | ||||
XM_011532905.1 | Intron | XP_011531207.1 | ||||
XM_011532906.1 | Intron | XP_011531208.1 | ||||
XM_017004325.1 | Intron | XP_016859814.1 | ||||
XM_017004326.1 | Intron | XP_016859815.1 | ||||
XM_017004327.1 | Intron | XP_016859816.1 | ||||
XM_017004328.1 | Intron | XP_016859817.1 |
Set Membership: |
HapMap |