Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCTGGCTGCTTCGTAGACATGC[C/T]CTAGAAAAAAAGGAGGAATCATGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300694 MIM: 300353 | ||||||||||||||||||||
Literature Links: |
MIR223 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR223 - microRNA 223 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VSIG4 - V-set and immunoglobulin domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100431.1 | 1068 | Missense Mutation | GAG,GGG | E,G 220 | NP_001093901.1 | |
NM_001184830.1 | 1068 | Missense Mutation | GAG,GGG | E,G 314 | NP_001171759.1 | |
NM_001184831.1 | 1068 | Missense Mutation | GAG,GGG | E,G 220 | NP_001171760.1 | |
NM_001257403.1 | 1068 | Missense Mutation | GAG,GGG | E,G 314 | NP_001244332.1 | |
NM_007268.2 | 1068 | Missense Mutation | GAG,GGG | E,G 314 | NP_009199.1 | |
XM_017029251.1 | 1068 | Missense Mutation | GAG,GGG | E,G 220 | XP_016884740.1 |