Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTGAGAGGCCATAGTGTATAGC[A/G]TTTGGGTGAAAAATGTGGGCTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612887 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEPT11 - septin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306147.1 | Intron | NP_001293076.1 | ||||
NM_018243.3 | Intron | NP_060713.1 | ||||
XM_006714259.3 | Intron | XP_006714322.1 | ||||
XM_017008400.1 | Intron | XP_016863889.1 | ||||
XM_017008401.1 | Intron | XP_016863890.1 | ||||
XM_017008402.1 | Intron | XP_016863891.1 | ||||
XM_017008403.1 | Intron | XP_016863892.1 | ||||
XM_017008404.1 | Intron | XP_016863893.1 |
Set Membership: |
HapMap |