Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGCAGAATCGGTATGATCCTT[C/T]TTCGGTGGCTTGACAACCTTAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602336 | ||||||||||||||||||||
Literature Links: |
ROR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ROR1 - receptor tyrosine kinase like orphan receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083592.1 | Intron | NP_001077061.1 | ||||
NM_005012.3 | Intron | NP_005003.2 | ||||
XM_011541526.1 | Intron | XP_011539828.1 | ||||
XM_017001376.1 | Intron | XP_016856865.1 | ||||
XM_017001377.1 | Intron | XP_016856866.1 |