Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATATAAAATGTTATATATTTAC[C/T]TGAAGATATAAGTTGGATAAACTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104613 MIM: 172480 MIM: 607940 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT6A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCT6A - chaperonin containing TCP1 subunit 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009186.1 | Intron | NP_001009186.1 | ||||
NM_001762.3 | Intron | NP_001753.1 |
PSPH - phosphoserine phosphatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA15 - small nucleolar RNA, H/ACA box 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUMF2 - sulfatase modifying factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |