Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCAGAGCAGGCTGGAACAGTGC[C/T]GTCTGGAAGGCTACAACAGCCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAZN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KAZN - kazrin, periplakin interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017999.2 | 4560 | Intron | NP_001017999.1 | |||
NM_001018000.3 | 4560 | Intron | NP_001018000.1 | |||
NM_001018001.2 | 4560 | Intron | NP_001018001.1 | |||
NM_015209.2 | 4560 | Intron | NP_056024.1 | |||
NM_201628.2 | 4560 | Missense Mutation | CGT,TGT | R,C 763 | NP_963922.2 | |
XM_005245795.4 | 4560 | Missense Mutation | CGT,TGT | R,C 861 | XP_005245852.1 | |
XM_011541074.2 | 4560 | UTR 3 | XP_011539376.1 | |||
XM_011541075.2 | 4560 | UTR 3 | XP_011539377.1 | |||
XM_011541076.2 | 4560 | UTR 3 | XP_011539378.1 | |||
XM_011541077.2 | 4560 | UTR 3 | XP_011539379.1 | |||
XM_011541080.2 | 4560 | Intron | XP_011539382.1 | |||
XM_011541081.2 | 4560 | UTR 3 | XP_011539383.1 | |||
XM_017000768.1 | 4560 | Intron | XP_016856257.1 | |||
XM_017000769.1 | 4560 | Intron | XP_016856258.1 | |||
XM_017000770.1 | 4560 | Intron | XP_016856259.1 | |||
XM_017000771.1 | 4560 | Intron | XP_016856260.1 | |||
XM_017000772.1 | 4560 | Intron | XP_016856261.1 |
TMEM51-AS1 - TMEM51 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |