Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCTGCCTCTCCCACCCGGGAGC[A/C]GCTGCCATACTCTGGCCTCAGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAB4 - GRB2 associated binding protein family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037814.1 | 2403 | UTR 3 | NP_001032903.1 | |||
XM_011546114.2 | 2403 | Intron | XP_011544416.1 | |||
XM_011546115.1 | 2403 | Intron | XP_011544417.1 | |||
XM_011546116.2 | 2403 | Intron | XP_011544418.1 | |||
XM_011546117.1 | 2403 | Intron | XP_011544419.1 | |||
XM_017028575.1 | 2403 | Intron | XP_016884064.1 | |||
XM_017028576.1 | 2403 | Intron | XP_016884065.1 |