Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGTGGTCACCTTTCATAGTGGT[A/G]AGTAGAGAATTAAAAGTGAGTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606414 MIM: 147796 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INSL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INSL6 - insulin like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007179.2 | Intron | NP_009110.2 | ||||
XM_011517702.2 | Intron | XP_011516004.1 |
JAK2 - Janus kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322194.1 | Intron | NP_001309123.1 | ||||
NM_001322195.1 | Intron | NP_001309124.1 | ||||
NM_001322196.1 | Intron | NP_001309125.1 | ||||
NM_001322198.1 | Intron | NP_001309127.1 | ||||
NM_001322199.1 | Intron | NP_001309128.1 | ||||
NM_001322204.1 | Intron | NP_001309133.1 | ||||
NM_004972.3 | Intron | NP_004963.1 |