Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATAGGAGAGAAAAGTTTGCCAAG[C/T]GTCTTCTATGGACTAAGCACTTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM246 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TMEM246 - transmembrane protein 246 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303107.1 | 2670 | UTR 3 | NP_001290036.1 | |||
NM_001303108.1 | 2670 | UTR 3 | NP_001290037.1 | |||
NM_032342.2 | 2670 | UTR 3 | NP_115718.1 | |||
XM_017015206.1 | 2670 | UTR 3 | XP_016870695.1 |
TMEM246-AS1 - TMEM246 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |