Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGACTTTTTCATATGCTAGAC[A/G]TCGTTTTTTGAATTGTGGTAGTTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf84 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C9orf84 - chromosome 9 open reading frame 84 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080551.2 | 4594 | Missense Mutation | CGT,TGT | R,C 1386 | NP_001074020.2 | |
NM_173521.4 | 4594 | Missense Mutation | CGT,TGT | R,C 1425 | NP_775792.4 | |
XM_006716974.2 | 4594 | Missense Mutation | CGT,TGT | R,C 1489 | XP_006717037.1 | |
XM_006716975.2 | 4594 | Missense Mutation | CGT,TGT | R,C 1454 | XP_006717038.1 | |
XM_011518302.2 | 4594 | Missense Mutation | CGT,TGT | R,C 1425 | XP_011516604.1 | |
XM_011518303.1 | 4594 | Missense Mutation | CGT,TGT | R,C 1425 | XP_011516605.1 | |
XM_011518306.1 | 4594 | Missense Mutation | CGT,TGT | R,C 1300 | XP_011516608.1 | |
XM_011518309.1 | 4594 | Intron | XP_011516611.1 | |||
XM_017014340.1 | 4594 | Missense Mutation | CGT,TGT | R,C 1376 | XP_016869829.1 | |
XM_017014341.1 | 4594 | Intron | XP_016869830.1 |