Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGGTCACTGCTGCTGATCAGAG[G/T]GCTGTAGTTTGGTTCAGGGCTACTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164012 MIM: 602327 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFKB2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NFKB2 - nuclear factor kappa B subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077494.3 | Intron | NP_001070962.1 | ||||
NM_001261403.2 | Intron | NP_001248332.1 | ||||
NM_001288724.1 | Intron | NP_001275653.1 | ||||
NM_001322934.1 | Intron | NP_001309863.1 | ||||
NM_001322935.1 | Intron | NP_001309864.1 | ||||
NM_002502.5 | Intron | NP_002493.3 | ||||
XM_011539830.2 | Intron | XP_011538132.1 | ||||
XM_011539831.2 | Intron | XP_011538133.1 | ||||
XM_017016278.1 | Intron | XP_016871767.1 |
PSD - pleckstrin and Sec7 domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |