Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCTGTCTCCTGGGCGGGTCCTTC[T/C]GATGTGAGTGCACTCCAGTGAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600106 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
INPP5A - inositol polyphosphate-5-phosphatase A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321042.1 | Intron | NP_001307971.1 | ||||
NM_005539.4 | Intron | NP_005530.3 | ||||
XM_017016204.1 | Intron | XP_016871693.1 | ||||
XM_017016205.1 | Intron | XP_016871694.1 |
LOC107984282 - uncharacterized LOC107984282 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |