Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGACTCACAGTCCAGTCCCAG[A/C]CCCTTCCTGAGCTCCTTTTGACCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607009 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPM6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRPM6 - transient receptor potential cation channel subfamily M member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177310.1 | Intron | NP_001170781.1 | ||||
NM_001177311.1 | Intron | NP_001170782.1 | ||||
NM_017662.4 | Intron | NP_060132.3 | ||||
XM_011518244.1 | Intron | XP_011516546.1 | ||||
XM_011518245.1 | Intron | XP_011516547.1 | ||||
XM_011518246.1 | Intron | XP_011516548.1 | ||||
XM_011518247.1 | Intron | XP_011516549.1 | ||||
XM_011518248.1 | Intron | XP_011516550.1 | ||||
XM_011518249.1 | Intron | XP_011516551.1 | ||||
XM_011518250.1 | Intron | XP_011516552.1 | ||||
XM_011518251.2 | Intron | XP_011516553.1 | ||||
XM_011518252.2 | Intron | XP_011516554.1 | ||||
XM_011518255.2 | Intron | XP_011516557.1 | ||||
XM_017014287.1 | Intron | XP_016869776.1 | ||||
XM_017014288.1 | Intron | XP_016869777.1 | ||||
XM_017014289.1 | Intron | XP_016869778.1 |
Set Membership: |
HapMap |