Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAACTATGAAGATTTTCTATGG[C/T]CCAGAAATTCAGGGCAGTTTGTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605673 MIM: 601206 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BTG4 - BTG anti-proliferation factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017589.3 | Intron | NP_060059.1 | ||||
XM_011542876.2 | Intron | XP_011541178.1 | ||||
XM_011542878.2 | Intron | XP_011541180.1 | ||||
XM_011542879.2 | Intron | XP_011541181.1 | ||||
XM_011542880.2 | Intron | XP_011541182.1 |
POU2AF1 - POU class 2 associating factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |