Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAGACCTCAAAGAAGGTCTTCT[C/T]AGCAATTGACAATGAATTTAACTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610079 MIM: 608621 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIAE PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SIAE - sialic acid acetylesterase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199922.1 | Intron | NP_001186851.1 | ||||
NM_170601.4 | Intron | NP_733746.1 | ||||
XM_017017930.1 | Intron | XP_016873419.1 |
SPA17 - sperm autoantigenic protein 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017425.3 | Intron | NP_059121.1 | ||||
XM_011542870.2 | Intron | XP_011541172.1 |
Set Membership: |
HapMap |