Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCTGCCTTCAACACCAGCCGCT[A/C]TCTGAGGGGAGCCCAGAGAGCCCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 159556 | ||||||||||||||||||||
Literature Links: |
MLLT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MLLT1 - MLLT1, super elongation complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005934.3 | 2067 | UTR 3 | NP_005925.2 | |||
XM_011528021.1 | 2067 | UTR 3 | XP_011526323.1 | |||
XM_011528022.1 | 2067 | UTR 3 | XP_011526324.1 | |||
XM_011528023.1 | 2067 | UTR 3 | XP_011526325.1 | |||
XM_017026819.1 | 2067 | UTR 3 | XP_016882308.1 |