Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTCAGAGCCCTCAAATGGCACG[A/G]AAGCGGCACCTGGTCTGCCAAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609061 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENAH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ENAH - enabled homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008493.1 | 10901 | UTR 3 | NP_001008493.1 | |||
NM_018212.4 | 10901 | UTR 3 | NP_060682.2 | |||
XM_005273187.2 | 10901 | UTR 3 | XP_005273244.1 | |||
XM_011544229.2 | 10901 | Intron | XP_011542531.2 | |||
XM_017001746.1 | 10901 | UTR 3 | XP_016857235.1 | |||
XM_017001747.1 | 10901 | Intron | XP_016857236.1 | |||
XM_017001748.1 | 10901 | Intron | XP_016857237.1 | |||
XM_017001749.1 | 10901 | Intron | XP_016857238.1 | |||
XM_017001750.1 | 10901 | Intron | XP_016857239.1 | |||
XM_017001751.1 | 10901 | UTR 3 | XP_016857240.1 | |||
XM_017001752.1 | 10901 | Intron | XP_016857241.1 | |||
XM_017001753.1 | 10901 | UTR 3 | XP_016857242.1 | |||
XM_017001754.1 | 10901 | UTR 3 | XP_016857243.1 | |||
XM_017001755.1 | 10901 | UTR 3 | XP_016857244.1 | |||
XM_017001756.1 | 10901 | UTR 3 | XP_016857245.1 | |||
XM_017001757.1 | 10901 | UTR 3 | XP_016857246.1 |
Set Membership: |
HapMap |