Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTGCTGGCTGTGTGGGCTTGGGA[A/T]AGCTACTTAACTTCTTTGTTCCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606498 | ||||||||||||||||||||
Literature Links: |
MS4A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MS4A3 - membrane spanning 4-domains A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031666.1 | Intron | NP_001026836.1 | ||||
NM_001031809.1 | Intron | NP_001026979.1 | ||||
NM_006138.4 | Intron | NP_006129.4 | ||||
XM_011545363.2 | Intron | XP_011543665.1 |
OOSP2 - oocyte secreted protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |