Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCATGGTCTCTGAAGATTATCT[C/T]GGCTCTGAAAAGCTTGGTTTAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613942 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SUN5 - Sad1 and UNC84 domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080675.3 | Intron | NP_542406.2 | ||||
XM_011528573.1 | Intron | XP_011526875.1 | ||||
XM_011528574.1 | Intron | XP_011526876.1 | ||||
XM_011528575.1 | Intron | XP_011526877.1 | ||||
XM_011528576.1 | Intron | XP_011526878.1 |
Set Membership: |
HapMap |