Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCCAGTCTCACCTGTGACCCAG[G/T]GCAAGTGGCAACACTTCTTGGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616320 MIM: 138760 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAHD1 - fumarylacetoacetate hydrolase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018104.2 | Intron | NP_001018114.1 | ||||
NM_001142398.1 | Intron | NP_001135870.1 | ||||
NM_031208.3 | Intron | NP_112485.1 |
HAGH - hydroxyacylglutathione hydrolase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MEIOB - meiosis specific with OB domains | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163560.2 | Intron | NP_001157032.1 | ||||
NM_152764.2 | Intron | NP_689977.2 |
Set Membership: |
HapMap |