Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTCTCCTCCAAGCACACCTATAT[C/T]ATGCCACTTCTTGTATCTCTCTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300655 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KLHL13 - kelch like family member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168299.1 | Intron | NP_001161771.1 | ||||
NM_001168300.1 | Intron | NP_001161772.1 | ||||
NM_001168301.1 | Intron | NP_001161773.1 | ||||
NM_001168302.1 | Intron | NP_001161774.1 | ||||
NM_001168303.1 | Intron | NP_001161775.1 | ||||
NM_033495.3 | Intron | NP_277030.2 | ||||
XM_011531409.2 | Intron | XP_011529711.1 | ||||
XM_011531410.2 | Intron | XP_011529712.1 | ||||
XM_011531411.1 | Intron | XP_011529713.1 | ||||
XM_017029950.1 | Intron | XP_016885439.1 |