Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGTATCTGGTTTACCAGTGGTG[A/G]TGGGAGGCTTCAGGACTGAAGGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615472 MIM: 613787 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPZ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COPZ1 - coatomer protein complex subunit zeta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271734.1 | 135 | Intron | NP_001258663.1 | |||
NM_001271735.1 | 135 | Intron | NP_001258664.1 | |||
NM_001271736.1 | 135 | Missense Mutation | ATG,GTG | M,V 1 | NP_001258665.1 | |
NM_016057.2 | 135 | Intron | NP_057141.1 | |||
XM_017018997.1 | 135 | Intron | XP_016874486.1 |
MIR148B - microRNA 148b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |