Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCCATTCTGTCTGGTGAGCAAT[A/G]TTCTGTGTTTTTTTGTTTGTTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610776 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DRAM1 - DNA damage regulated autophagy modulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018370.2 | Intron | NP_060840.2 | ||||
XM_005269004.2 | Intron | XP_005269061.1 | ||||
XM_005269005.2 | Intron | XP_005269062.1 | ||||
XM_017019578.1 | Intron | XP_016875067.1 |