Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCCATAAAGTTCCCTGGCACCC[C/T]TTTTTAGTCAATTTCACTCACGTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603282 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF204P PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF204P - zinc finger protein 204, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF391 - zinc finger protein 391 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076781.2 | Intron | NP_001070249.1 | ||||
NM_001322288.1 | Intron | NP_001309217.1 | ||||
NM_001322289.1 | Intron | NP_001309218.1 | ||||
NM_001322293.1 | Intron | NP_001309222.1 | ||||
XM_005249082.3 | Intron | XP_005249139.1 | ||||
XM_006715086.2 | Intron | XP_006715149.1 | ||||
XM_011514569.1 | Intron | XP_011512871.1 |
Set Membership: |
HapMap |