Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAACCCCACTAGTAATGGCAGA[A/G]CTCGAGGAAGGGGCTTGAACTCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LINC01055 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC01055 - long intergenic non-protein coding RNA 1055 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPERT - spermatid associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286341.1 | Intron | NP_001273270.1 | ||||
NM_001286342.1 | Intron | NP_001273271.1 | ||||
NM_152719.2 | Intron | NP_689932.1 | ||||
XM_011534971.2 | Intron | XP_011533273.1 |