Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGATTAGATTAGACCAAGCAGG[G/T]TAATGCAGCGTAATCTCCCATCTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601774 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGAT5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MGAT5 - mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002410.4 | Intron | NP_002401.1 | ||||
XM_005263668.4 | Intron | XP_005263725.1 | ||||
XM_005263669.4 | Intron | XP_005263726.1 | ||||
XM_005263670.3 | Intron | XP_005263727.1 | ||||
XM_006712534.3 | Intron | XP_006712597.1 | ||||
XM_011511199.2 | Intron | XP_011509501.1 | ||||
XM_011511200.1 | Intron | XP_011509502.1 | ||||
XM_011511201.2 | Intron | XP_011509503.1 | ||||
XM_011511202.1 | Intron | XP_011509504.1 | ||||
XM_011511204.2 | Intron | XP_011509506.1 | ||||
XM_017004147.1 | Intron | XP_016859636.1 | ||||
XM_017004148.1 | Intron | XP_016859637.1 | ||||
XM_017004149.1 | Intron | XP_016859638.1 |
MIR3679 - microRNA 3679 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |