Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604933 MIM: 604746 MIM: 613931 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUTYH PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TESK2 - testis-specific kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOE1 - target of EGR1, member 1 (nuclear) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025077.3 | 398 | Intron | NP_079353.3 | |||
XM_005270412.3 | 398 | Intron | XP_005270469.1 | |||
XM_005270413.4 | 398 | UTR 5 | XP_005270470.1 | |||
XM_011540569.2 | 398 | Intron | XP_011538871.1 |
Set Membership: |
HapMap |